Merck
  • Cre-mediated cell ablation contests mast cell contribution in models of antibody- and T cell-mediated autoimmunity.
Cre-mediated cell ablation contests mast cell contribution in models of antibody- and T cell-mediated autoimmunity.
Immunity (2011-11-22)
Thorsten B Feyerabend, Anne Weiser, Annette Tietz, Michael Stassen, Nicola Harris, Manfred Kopf, Peter Radermacher, Peter Möller, Christophe Benoist, Diane Mathis, Hans Jörg Fehling, Hans-Reimer Rodewald
ABSTRACT
Immunological functions of mast cells remain poorly understood. Studies in Kit mutant mice suggest key roles for mast cells in certain antibody- and T cell-mediated autoimmune diseases. However, Kit mutations affect multiple cell types of both immune and nonimmune origin. Here, we show that targeted insertion of Cre-recombinase into the mast cell carboxypeptidase A3 locus deleted mast cells in connective and mucosal tissues by a genotoxic Trp53-dependent mechanism. Cre-mediated mast cell eradication (Cre-Master) mice had, with the exception of a lack of mast cells and reduced basophils, a normal immune system. Cre-Master mice were refractory to IgE-mediated anaphylaxis, and this defect was rescued by mast cell reconstitution. This mast cell-deficient strain was fully susceptible to antibody-induced autoimmune arthritis and to experimental autoimmune encephalomyelitis. Differences comparing Kit mutant mast cell deficiency models to selectively mast cell-deficient mice call for a systematic re-evaluation of immunological functions of mast cells beyond allergy.