Merck
  • One-step enzymatic synthesis of nucleosides from low water-soluble purine bases in non-conventional media.
One-step enzymatic synthesis of nucleosides from low water-soluble purine bases in non-conventional media.
Bioresource technology (2011-12-27)
Jesús Fernández-Lucas, Alba Fresco-Taboada, Isabel de la Mata, Miguel Arroyo
ABSTRACT
The effect of several water-miscible cosolvents on activity and stability of soluble and immobilized 2'-deoxyribosyltransferase from Lactobacillus reuteri on Sepabeads® has been studied in order to establish optimal conditions for enzymatic synthesis of nucleosides using purine bases with low solubility in aqueous buffer. As a rule of thumb, there was a general reduction of soluble enzyme activity when cosolvent content was gradually increased in reaction medium. In contrast, immobilized enzyme activity was enhanced 1.2-1.4-fold at 20% of methanol, ethanol, 2-propanol, diethylene glycol, and acetone; and at 10% and 30% acetonitrile. Likewise, highest increased activity (1.8-fold) was also obtained in presence of 20% acetonitrile. Immobilized enzyme was successfully used in the synthesis of 2'-deoxyxanthosine and 2'-deoxyguanosine using 2'-deoxyuridine as sugar donor and the corresponding poor water-soluble base in the presence of 30% of methanol, ethanol, 2-propanol, ethylene glycol, acetonitrile, and DMSO, giving high nucleoside yields at 4h.