Merck
  • Structure-activity relationship study of selective benzimidazole-based inhibitors of Cryptosporidium parvum IMPDH.
Structure-activity relationship study of selective benzimidazole-based inhibitors of Cryptosporidium parvum IMPDH.
Bioorganic & medicinal chemistry letters (2012-02-09)
Sivapriya Kirubakaran, Suresh Kumar Gorla, Lisa Sharling, Minjia Zhang, Xiaoping Liu, Soumya S Ray, Iain S Macpherson, Boris Striepen, Lizbeth Hedstrom, Gregory D Cuny
ABSTRACT
Cryptosporidium parasites are important waterborne pathogens of both humans and animals. The Cryptosporidium parvum and Cryptosporidium hominis genomes indicate that the only route to guanine nucleotides is via inosine 5'-monophosphate dehydrogenase (IMPDH). Thus the inhibition of the parasite IMPDH presents a potential strategy for treating Cryptosporidium infections. A selective benzimidazole-based inhibitor of C. parvum IMPDH (CpIMPDH) was previously identified in a high throughput screen. Here we report a structure-activity relationship study of benzimidazole-based compounds that resulted in potent and selective inhibitors of CpIMPDH. Several compounds display potent antiparasitic activity in vitro.