- Immunogold-silver staining: new method of immunostaining with enhanced sensitivity.
Immunogold-silver staining: new method of immunostaining with enhanced sensitivity.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1983-07-01)
C S Holgate, P Jackson, P N Cowen, C C Bird
PMID6189883
ABSTRACT
A new method for demonstrating antigens in paraffin sections of formol sublimate-fixed tissue is described that utilizes an "indirect" immunohistological technique employing immunoglobulin adsorbed to colloidal gold as the secondary antiserum. The gold particles introduced to antigenic sites are revealed by a silver precipitation reaction. This technique, the immunogold-silver staining method, is of much enhanced sensitivity (up to 200-fold) as compared with standard immunoperoxidase and immunogold staining methods. The results have been confirmed in a study of immunoglobulins in reactive human tonsil. The use of this new method for double immunolabeling is also described.