- IRAK (Pelle) family member IRAK-2 and MyD88 as proximal mediators of IL-1 signaling.
IRAK (Pelle) family member IRAK-2 and MyD88 as proximal mediators of IL-1 signaling.
Science (New York, N.Y.) (1997-12-31)
M Muzio, J Ni, P Feng, V M Dixit
PMID9374458
ABSTRACT
The interleukin-1 receptor (IL-1R) signaling pathway leads to nuclear factor kappa B (NF-kappaB) activation in mammals and is similar to the Toll pathway in Drosophila: the IL-1R-associated kinase (IRAK) is homologous to Pelle. Two additional proximal mediators were identified that are required for IL-1R-induced NF-kappaB activation: IRAK-2, a Pelle family member, and MyD88, a death domain-containing adapter molecule. Both associate with the IL-1R signaling complex. Dominant negative forms of either attenuate IL-1R-mediated NF-kappaB activation. Therefore, IRAK-2 and MyD88 may provide additional therapeutic targets for inhibiting IL-1-induced inflammation.