- Comparison of two methods of preparing enzyme-antibody conjugates: application of these conjugates for enzyme immunoassay.
Comparison of two methods of preparing enzyme-antibody conjugates: application of these conjugates for enzyme immunoassay.
Analytical biochemistry (1979-11-15)
M J O'Sullivan, E Gnemmi, D Morris, G Chieregatti, A D Simmonds, M Simmons, J W Bridges, V Marks
PMID120684