Merck
  • Synthesis and resolution of racemic eliprodil and evaluation of the enantiomers of eliprodil as NMDA receptor antagonists.
Synthesis and resolution of racemic eliprodil and evaluation of the enantiomers of eliprodil as NMDA receptor antagonists.
Bioorganic & medicinal chemistry letters (2000-07-13)
J Pabel, G Höfner, K T Wanner
ABSTRACT
A short synthesis of the NMDA receptor antagonist (rac)-Eliprodil (9) and its resolution into the enantiomers by chiral HPLC is described. The enantiomers (R)-9 and (S)-9 were found to exhibit markedly different affinities for NR2B subunit containing NMDA receptors.