Merck
  • Improved water solubility of neohesperidin dihydrochalcone in sweetener blends.
Improved water solubility of neohesperidin dihydrochalcone in sweetener blends.
Journal of agricultural and food chemistry (2001-02-15)
O Benavente-García, J Castillo, M J Del Baño, J Lorente
ABSTRACT
Significant technological advantages in terms of sweetness synergy and hence cost-saving can be obtained if neohesperidin dihydrochalcone (NHDC) is used in sweetener blends with other intense or bulk sweeteners. The combination of NHDC with sodium saccharin or sodium cyclamate is an excellent method to improve the water solubility at room temperature of NHDC. In the case of NHDC-sodium saccharin, two different methods for blend preparation, a simple mixture and a cosolubilized mixture, can be used, with similar results obtained for solubility and solution stability properties. To improve temporally the water solubility of NHDC in combination with sodium cyclamate, it is absolutely necessary to prepare cosolubilized blends.
Sigma-Aldrich
Neohesperidin dihydrochalcone, ≥96%, FG
Sigma-Aldrich
Neohesperidin dihydrochalcone, ≥95% (HPLC)