Merck
  • Two chromone-secoiridoid glycosides and three indole alkaloid glycosides from Neonauclea sessilifolia.
Two chromone-secoiridoid glycosides and three indole alkaloid glycosides from Neonauclea sessilifolia.
Phytochemistry (2003-03-07)
Atsuko Itoh, Takao Tanahashi, Naotaka Nagakura, Toyoyuki Nishi
ABSTRACT
From the dried roots of Neonauclea sessilifolia, two new chromone-secoiridoid glycosides, sessilifoside and 7"-O-beta-D-glucopyranosylsessilifoside, and three novel indole alkaloid glycosides, neonaucleosides A, B, and C, were isolated along with the main known glycosides, 5-hydroxy-2-methylchromone-7-O-beta-D-apiofuranosyl-(1-->6)-beta-D-glucopyranoside, sweroside, loganin, grandifloroside, and quinovic acid 3 beta-O-beta-D-quinovopyranoside-28-O-beta-D-glucopyranoside. The structures of these new glycosides were determined by spectroscopic and chemical means. Neonaucleoside A and its C-3 epimer were prepared from secologanin and tryptamine.