Merck
  • Increased concentrations of histamine and its metabolite, tele-methylhistamine and down-regulation of histamine H3 receptor sites in autopsied brain tissue from cirrhotic patients who died in hepatic coma.
Increased concentrations of histamine and its metabolite, tele-methylhistamine and down-regulation of histamine H3 receptor sites in autopsied brain tissue from cirrhotic patients who died in hepatic coma.
Journal of hepatology (2003-09-16)
Violina Lozeva, Leena Tuomisto, Juhani Tarhanen, Roger F Butterworth
ABSTRACT
Hepatic encephalopathy (HE) is a serious neuropsychiatric complication of chronic liver disease. To determine whether changes in the central histaminergic system are a feature of human HE, we studied histamine, tele-methylhistamine, and presynaptic autoregulatory H(3) receptors in cerebral cortex and caudate-putamen obtained at autopsy from six cirrhotic patients and six appropriately matched controls. Histamine was assayed by HPLC; tele-methylhistamine by GC-MS. H(3) receptors were studied by in vitro receptor binding using [3H]R-alpha-methylhistamine as ligand. In HE patients, there was a significant fourfold increase of histamine in caudate-putamen and a significant increase in all cortical regions studied. tele-Methyhistamine was also increased and the densities of histamine H(3) receptor sites were significantly decreased in patient material. These findings are consistent with activation of the histaminergic system in HE. Given that histamine participates in the regulation of arousal and circadian rhythmicity, they indicate that induction of central histamine mechanisms may contribute to the development of neuropsychiatric symptoms, such as sleep disturbances and altered circadian rhythms in chronic HE and suggest that pharmacological manipulation of the histaminergic system could be beneficial in the treatment of HE in chronic liver failure.