Merck
Mercapturic acid formation from 2-aminopyrazine in rats.
Xenobiotica; the fate of foreign compounds in biological systems (1992-11-01)
W H Lunn, J K Shadle
ABSTRACT
1. The compound 2-aminopyrazine was given by oral gavage to normal rats and their urine collected. 2. A mercapturic acid containing the 2-aminopyrazine moiety was isolated from this urine. This represents the first example of this type of a metabolite from a pyrazine.