Merck
  • A microELISA assay for detection of anti-HLA activity of mouse monoclonal antibodies using an Astroscan 2100 automated plate reader.
A microELISA assay for detection of anti-HLA activity of mouse monoclonal antibodies using an Astroscan 2100 automated plate reader.
Journal of immunological methods (1992-04-27)
A M Sadler, P Krausa, S G Marsh, J M Heyes, J G Bodmer
ABSTRACT
A microenzyme-linked immunosorbent assay (microELISA) method has been developed using an Astroscan 2100 system automated plate reader which was initially designed for tissue typing by a two colour fluorescent microcytotoxicity assay. A 96-well plate ELISA used for screening mouse monoclonal antibodies raised against surface HLA antigens has been modified for use with the Astroscan plate reader and 72-well typing trays. The existing substrate 4-methylumbelliferyl-beta-D-galactoside (4MUG) has been replaced with fluorescein-di-beta-D-galactopyranoside (FDG), to provide a wavelength (530 nm) detectable by the Astroscan or other automated plate readers designed for reading microcytotoxicity assay plates. The assay volumes have also been reduced tenfold for use with Terasaki microtest plates. The assay now has the major advantage of requiring only 5 microliters of test supernatant allowing hybridomas to be screened earlier during a fusion and on a wider cell panel. The use of the large panel which includes B lymphoblastoid cell lines (B-LCL) and mouse L cell transfectants expressing HLA genes, reduces the length of time the hybridomas need to be kept in tissue culture before selection. Other advantages include the reduction in the number of target cells required, smaller volumes of reagents throughout the assay and the ability to screen cytotoxic as well as non-cytotoxic monoclonal antibodies. The sensitivity of this microELISA proved to be comparable with the original assay and so provides an efficient screening method for monoclonal antibodies.