Merck
  • A case study on fuel oil contamination in a mangrove swamp in Hong Kong.
A case study on fuel oil contamination in a mangrove swamp in Hong Kong.
Marine pollution bulletin (2005-07-19)
Nora F Y Tam, Teresa W Y Wong, Y S Wong
ABSTRACT
Mangroves commonly found along tropical and subtropical coastlines are susceptible to oil pollution. In December 2000, around 500 1 m tall Kandelia candel saplings at the age of 3-5 years old located at the foreshore region of Sheung Pak Nai swamp, Hong Kong SAR, were found to be damaged by oil pollution. More than 80% of the saplings were either dead or washed away and leaving less than 5% healthy saplings with dense green leaves. Elevated concentrations of light n-alkanes (ranging from n-C14 to n-C20), pristane and phytane were recorded in surface sediments collected in December 2000. The ratio between light and total n-alkanes was 0.4. The total petroleum hydrocarbons (60-80 microg g(-1) TPH) and unresolved complex mixtures (60-70 microg g(-1) UCM) were higher than the background values of other mangrove sediments in Hong Kong, which were 40 and 20 microg g(-1), respectively. In certain root zone sediments, TPH concentrations were above 1000 microg g(-1). These results suggest that surface sediments in Sheung Pak Nai were contaminated by petroleum oil, most likely by illegal discharge of fuel oil which occurred between 1998 and 2002. One year later, in December 2001, unhealthy saplings had recovered and re-grown. The concentrations of TPH and UCM in sediments declined to around 40 microg g(-1), pristane and phytane dropped by 80%, and the ratio of light to total n-alkanes was 0.15, suggesting that residual oil in sediments was weathered leading to a remarkable recovery of the unhealthy saplings.