Merck
  • Fabrication of beta-Ni(OH)2 and NiO hollow spheres by a facile template-free process.
Fabrication of beta-Ni(OH)2 and NiO hollow spheres by a facile template-free process.
Chemical communications (Cambridge, England) (2005-10-18)
Yong Wang, Qingshan Zhu, Huigang Zhang
ABSTRACT
Ni(OH)2 hollow microspheres with beta-Ni(OH)2 nanosheets as the in situ formed building units were fabricated via a novel template-free approach in a strong alkaline solution of glycine, and can be converted into NiO hollow microspheres by a thermal decomposition process.