Merck
  • Mechanism of constipation in familial amyloid polyneuropathy: a case report.
Mechanism of constipation in familial amyloid polyneuropathy: a case report.
Internal medicine (Tokyo, Japan) (2006-11-16)
Takashi Ito, Ryuji Sakakibara, Shoichi Ito, Tomoyuki Uchiyama, Zhi Liu, Tatsuya Yamamoto, Taketo Yamaguchi, Takeo Odaka, Morihiro Higashi, Takamichi Hattori
ABSTRACT
Previously no functional study has been available for the mechanism of constipation in familial amyloid polyneuropathy (FAP). We performed a gut function test in a 78-year-old woman with transthyretin-type FAP who had severe constipation. The gut function test showed a prolonged colonic transit time, a low anal pressure at rest and upon squeezing, loss of the spontaneous phasic rectal contractions, and weak abdominal strain. All these abnormalities suggested a peripheral autonomic dysfunction, corresponding to the enteric neuropathy that was confirmed upon autopsy.