Merck
  • Indirect enantioresolution of (R,S)-mexiletine by reversed-phase high-performance liquid chromatography via diastereomerization with [(S,S)-O,O'-di-p-toluoyl tartaric acid anhydride], (S)-naproxen and nine chiral reagents synthesized as variants of Marfey's reagent.
Indirect enantioresolution of (R,S)-mexiletine by reversed-phase high-performance liquid chromatography via diastereomerization with [(S,S)-O,O'-di-p-toluoyl tartaric acid anhydride], (S)-naproxen and nine chiral reagents synthesized as variants of Marfey's reagent.
Biomedical chromatography : BMC (2010-06-30)
R Bhushan, Shivani Tanwar, Shuchi Dixit
ABSTRACT
Eleven chiral derivatizing reagents (CDRs) were used for preparation of diastereomers of (R,S)-mexiletine containing a primary amino group in close proximity to the stereogenic center. One anhydride, namely [(S,S)-O,O'-di-p-toluoyl tartaric acid anhydride] was synthesized and (S)-naproxen was used as such as the chiral derivatizing reagent. The other nine CDRs were synthesized by substituting one of the fluorine atoms in 1,5-difluoro-2,4-dinitrobenzene with six amino acid amides and three amino acids. The diastereomers were separated by reversed-phase high-performance liquid chromatography. The method was validated for linearity, accuracy, limit of detection and limit of quantification. The limit of detection was found in the range of 10-30  pmol.
Product Description
Supelco
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, derivatization grade (chiral), LiChropur, ≥99.0%
Sigma-Aldrich
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, powder