Merck
[Aflatoxins--health risk factors].
Bacteriologia, virusologia, parazitologia, epidemiologia (Bucharest, Romania : 1990) (2010-11-03)
Nicoleta Manuela Miliţă, Gr Mihăescu, Carmen Chifiriuc
ABSTRACT
Aflatoxins are secondary metabolites produced by a group of strains, mainly Aspergillus and Penicillium species. These mycotoxins are bifurano-coumarin derivatives group with four major products B1, B2, G1 and G2 according to blue or green fluorescence emitted in ultraviolet light and according to chromatographic separation. After metabolism of aflatoxin B1 and B2 in the mammalian body, result two metabolites M1 and M2 as hydroxylated derivatives of the parent compound. Aflatoxins have high carcinogenic potential, the most powerful carcinogens in different species of animals and humans. International Agency for Research on Cancer has classified aflatoxin B1 in Group I carcinogens. The target organ for aflatoxins is the liver. In chronic poisoning, aflatoxin is a risk to health, for a long term causing cancer (hepatocellular carcinoma), and in acute intoxications aflatoxin is lethal. This work purpose to discuss aflatoxins issue: the synthesis, absorption and elimination of aflatoxins, the toxicity mechanisms, and measures to limit the content of aflatoxins in food