Merck
  • Metal-dependent and redox-selective coordination behaviors of metalloligand [MoV(1,2-benzenedithiolato)3]- with CuI/AgI ions.
Metal-dependent and redox-selective coordination behaviors of metalloligand [MoV(1,2-benzenedithiolato)3]- with CuI/AgI ions.
Inorganic chemistry (2011-03-11)
Takeshi Matsumoto, Ho-Chol Chang, Atsushi Kobayashi, Kohei Uosaki, Masako Kato
ABSTRACT
The synthesis and characterization of two coordination polymers, {Cu(I)[Mo(V)(bdt)(3)]·0.5Et(2)O}(n) (1·0.5Et(2)O, bdt: o-benzenedithiolato) and {Ag(I)[Mo(V)(bdt)(3)]}(n) (2), composed of redox-active [Mo(V)(bdt)(3)](-) metalloligand with Cu(I) and Ag(I) ions are reported. The complexation reactions of [Mo(V)(bdt)(3)](-) with Cu(II)(ClO(4))(2) or Ag(I)ClO(4) commonly lead to the formation of one-dimensional (1-D) coordination polymers. The presence of Cu(I) in 1·0.5Et(2)O strongly indicates that the Cu(II) ion is reduced during the complexation reaction with [Mo(V)(bdt)(3)](-), which acts as an electron donor. The total dimensionalities of the assembled structures of 1·0.5Et(2)O and 2 are significantly different and related to the type of additional metal ions, Cu(I) and Ag(I). In contrast to the isolated 1-D chain structure of 1·0.5Et(2)O, complex 2 has a three-dimensional (3-D) assembled structure constructed from additional π-π stacking interactions between adjacent [Mo(V)(bdt)(3)](-) moieties. These structural differences influence the solubility of the complexes in organic solvents; complex 1·0.5Et(2)O is soluble as origomeric species in highly polar solvents, while 2 is insoluble in organic solvents and water. Coordination polymers 1·0.5Et(2)O and 2 were investigated by UV-vis spectroscopy in the solid state, and that in solution together with their electrochemical properties were also investigated for 1 because of its higher solubility in polar organic solvents. Complex 1·0.5Et(2)O dissolved in CH(3)CN demonstrates concentration-dependent UV-vis spectra supporting the presence of coordinative interactions between [Mo(V)(bdt)(3)](-) moieties and Cu(I) ions to create the origomeric species even in solutions, an observation that is supported also by electrochemical experiments.