Merck
  • [The protein expression profiles induced by trimethyltin chloride in Vero cells].
[The protein expression profiles induced by trimethyltin chloride in Vero cells].
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases (2012-02-24)
Yun Xiao, Li-jin Zhu, Li Jv, Ya-ling Qian, Xing Zhang
ABSTRACT
To explore the biomarkers and mechanism of kidney toxicity induced by trimethyltin chloride (TMT-Cl) through analyzing the differences of protein expression profiles between vero cells and vero cells exposed to TMT-Cl. The differences of protein expression levels of three paired samples of vero cells and vero cells exposed to TMT-Cl were compared by two-dimensional gel electrophoresis (2-DE) and liquid chromatography-electrospray ionization-linear trap quadrupole (LC-ESI-LTQ). The differences of expression levels of Annexin A1 and α-Tubulin proteins were validated with western blot assay, and the differences of mRNA expression levels of Annexin A1 and α-Tubulin genes were detected with quantitative reverse transcriptase-polymerase chain reaction (qRT-PCR). Fifteen spots of differential expression in protein profiles between vero cells and vero cells exposed to TMT-Cl were found, and 9 of these spots were identified by LC-ESI-LTQ. The expression levels of 3 proteins (Annexin A1,similar to RAN protein and a hypothetical protein) increased and the expression levels of 6 proteins(growth factor receptor-bound protein 10, tubulin alpha 6, heterogeneous nuclear ribonucleoprotein, similar to elongation factor SIII p15 subunit, S-adenosylhomocysteine hydrolase and a hypothetical protein) reduced. The expression levels of α-Tubulin protein and mRNA significantly decreased in vero cells exposed to TMT-Cl, as compared with vero cells (P < 0.01). The expression of Annexin A1 protein in all exposure groups was significantly up-regulated, the expression of Annexin A1 mRNA in the groups exposed to 25 and 50 µmol/L TMT-Cl was significantly down-regulated, and The expression of Annexin A1 mRNA in the group exposed to 100 µmol/L TMT-Cl was significantly up-regulated (P < 0.01). The results of present study suggest that 9 proteins with differential expression detected by LC-ESI-LTQ may be related to the kidney toxicity induced by TMT-Cl, which can serve as the biomarkers of early diagnosis and therapeutic effect for the kidney toxicity induced by TMT-Cl.