Merck
  • Oxidative stress and antioxidant response in Hypericum perforatum L. plants subjected to low temperature treatment.
Oxidative stress and antioxidant response in Hypericum perforatum L. plants subjected to low temperature treatment.
Journal of plant physiology (2012-05-12)
Matúš Skyba, Linda Petijová, Ján Košuth, Dimitrina Petrova Koleva, Tsveta Gancheva Ganeva, Veneta Mihova Kapchina-Toteva, Eva Cellárová
ABSTRACT
Extreme low temperatures cause plants multiple stresses, among which oxidative stress is presumed to be the major component affecting the resultant recovery rate. Plants of Hypericum perforatum L., which are known especially for the photodynamic activities of hypericins capable of producing reactive oxygen species under exposure to visible light, were observed to display a substantial increase and persistence in active oxygen production at least two months after recovery from cryogenic treatment. In an effort to uncover the causative mechanism, the individual contributions of wounding during explant isolation, dehydration and cold were examined by means of antioxidant profiling. The investigation revealed activation of genes coding for enzymatic antioxidant catalase and superoxide dismutase at both the transcript and protein levels. Interestingly, plants responded more to wounding than to either low-temperature associated stressor, presumably due to tissue damage. Furthermore, superoxide dismutase zymograms showed the Cu/Zn isoforms as the most responsive, directing the ROS production particularly to chloroplasts. Transmission electron microscopy revealed chloroplasts as damaged structures with substantial thylakoid ruptures.
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%