Merck
  • Bystander effect in tumor cells produced by Iodine-125 labeled human lymphocytes.
Bystander effect in tumor cells produced by Iodine-125 labeled human lymphocytes.
International journal of radiation biology (2012-06-21)
Omar Mamlouk, Pichumani Balagurumoorthy, Ketai Wang, S James Adelstein, Amin I Kassis
ABSTRACT
To investigate the ability of human lymphocytes labeled with DNA-incorporated (125)I to exert an inhibitory (antiproliferative) bystander effect on co-cultured human colon adenocarcinoma LS174T cells in vitro. Human peripheral blood lymphocytes were stimulated to synthesize DNA in the presence of phytohemagglutinin (PHA) and labeled with 5-[(125)I]iodo-2'-deoxyuridine. Human colon adenocarcinoma LS174T cells were co-cultured with the (125)I-labeled lymphocytes in various ratios for 5 days and the proliferation of the LS174T cells was assessed. Further, the supernatant media from these co-cultures were: (i) Transferred to LS174T cells and their proliferation measured after 5 days, (ii) used to assess the clonogenic survival of LS174T cells, and (iii) screened for factors that suppress growth. A significant reduction in the proliferation of LS174T cells was observed when co-cultured either with (125)I-labeled lymphocytes (56 ± 3.5%) or the supernatant media (52.5 ± 1.3%) obtained from these co-cultures. Clonogenic survival of LS174T cells grown in the supernatant media corroborated the decrease in tumor cell growth. The observed reduction in the proliferation of LS174T cells in presence of (125)I-labeled lymphocytes or media obtained from such co-cultures can be attributed to an inhibitory (antiproliferative) bystander effect, probably mediated by factor(s) released from the dying (125)I-labeled lymphocytes.