- [Interaction of active chlortriazine dyes with blood proteins].
[Interaction of active chlortriazine dyes with blood proteins].
Gigiena i sanitariia (1990-08-01)
E L Balabanova, B A Kurliandskiĭ, N P Parinova
PMID2283067
ABSTRACT
The study was undertaken to analyze the capacity of organic active chlorotriazine pigments to make contact with blood serum proteins. It was established that pigment toxicity was inversely correlated to the capacity of making contact with blood serum proteins (r = 99%) and directly correlated to the capacity of making contact with albumin (r = 96%).