- 2D difference gel electrophoresis reference map of a Fusarium graminearum nivalenol producing strain.
2D difference gel electrophoresis reference map of a Fusarium graminearum nivalenol producing strain.
Electrophoresis (2012-11-23)
Matias Pasquali, Tommaso Serchi, Jenny Renaut, Lucien Hoffmann, Torsten Bohn
PMID23172383
ABSTRACT
Fusarium graminearum is widely studied as a model for toxin production among plant pathogenic fungi. A 2D DIGE reference map for the nivalenol-producing strain 453 was established. Based on a whole protein extract, all reproducible spots were systematically picked and analyzed by MALDI-TOF/TOF, leading to the identification of 1102 protein species. The obtained map contributes to the annotation of the genome by identifying previously nondescribed hypothetical proteins and will serve as a reference for future studies aiming at deciphering F. graminearum biology and chemotype diversity.
Product Description
Nivalenol in acetonitrile, IRMM®, certified reference material