Merck
  • Surgical treatment of ruptured anterior circulation aneurysms: comparison of pterional and supraorbital keyhole approaches.
Surgical treatment of ruptured anterior circulation aneurysms: comparison of pterional and supraorbital keyhole approaches.
Neurosurgery (2012-12-05)
Nohra Chalouhi, Pascal Jabbour, Ibrahim Ibrahim, Robert M Starke, Philippe Younes, Gilles El Hage, Elie Samaha
ABSTRACT
Recent advancements in microsurgical techniques and instrumentation have allowed the development of the keyhole approach in aneurysm surgery. To compare the safety, efficacy, and 1-year clinical outcome of supraorbital keyhole and standard pterional approaches for ruptured anterior circulation aneurysms. A total of 87 patients underwent surgical clipping, 40 through the pterional and 47 through the supraorbital keyhole approach. Baseline demographics, operative time, procedural complications, and 1-year patient outcome were retrospectively compared. The 2 groups were comparable with respect to baseline characteristics, with the exception of a higher proportion of small aneurysms (<7 mm) in the supraorbital group (70.2% vs. 37.5%, P = .002). Total operative time was significantly shorter in the supraorbital group (205 minutes, P < .001) compared with the pterional group (256 minutes). The rate of procedural complications was lower in patients treated through the pterional (17.5%) vs the supraorbital approach (23.4%, P = .4). Intraoperative aneurysm ruptures occurred more frequently in the supraorbital group (10.6% vs. 2.5%). No patient experienced early or late rebleeding in either group. One year after treatment, 75% (30/40) of patients achieved a favorable outcome (Glasgow outcome scale IV or V) in the pterional group vs 76.6% (36/47) in the supraorbital group (P = .8). The rate of procedural complications may be higher with the supraorbital keyhole approach, but overall patient outcomes appear to be comparable. The pterional approach is a simple, reliable, and efficient procedure. The keyhole approach may be an acceptable alternative for neurosurgeons who have gained sufficient experience with the technique, especially for small noncomplex aneurysms.