Merck
  • Anti-hyperalgesic activity of corilagin, a tannin isolated from Phyllanthus niruri L. (Euphorbiaceae).
Anti-hyperalgesic activity of corilagin, a tannin isolated from Phyllanthus niruri L. (Euphorbiaceae).
Journal of ethnopharmacology (2013-01-22)
Jeverson Moreira, Luiz Carlos Klein-Júnior, Valdir Cechinel Filho, Fátima de Campos Buzzi
ABSTRACT
Corilagin (β-1-O-galloyl-3,6-(R)-hexahydroxydiphenoyl-D-glucose) is a tannin isolated from Phyllanthus niruri (Euphorbiaceae). This plant is well known for their therapeutic purposes to treat several diseases associated with dolorous process and are used in several ethno-medicines in tropical and subtropical countries. This study was designed to evaluate the anti-hyperalgesic activity of corilagin using chemically and thermally based nociception models in mice. Corilagin was isolated from Phyllanthus niruri (Euphorbiaceae) by extraction and chromatographic procedures and the anti-hyperalgesic activity was evaluated by using writhing, formalin, capsaicin, glutamate and hot plate tests in mice. Corilagin presented activity in acetic acid model with the ID50 calculated value of 6.46 (3.09-13.51) being about 20.6 fold more potent than acetylsalicylic acid. It also exhibited activity against the first phase of formalin test with ID50 value of 18.38 (15.15-22.59) μmol/kg. In the capsaicin and glutamate models, corilagin demonstrated significant activity at the 3 mg/kg. The experimental data demonstrated that corilagin exhibits anti-hyperalgesic activity that may be due to interaction with the glutamatergic system.