Merck
  • Occurrence and distribution of antifouling biocide Irgarol-1051 in coastal waters of Peninsular Malaysia.
Occurrence and distribution of antifouling biocide Irgarol-1051 in coastal waters of Peninsular Malaysia.
Marine pollution bulletin (2013-03-16)
Hassan Rashid Ali, Marinah Mohd Arifin, Mohammed Ali Sheikh, Noor Azhar Mohamed Shazili, Zainudin Bachok
ABSTRACT
Emerging booster biocides contamination raises particular attention in the marine ecosystem health. This study provides the baseline data on the occurrence of Irgarol-1051 (2-methylthio-4-tert-butylamino-6-cyclopropylamiono-s-triazine) in the selected coastal water around Malaysia. The maximum detected concentration of Irgarol was 2021 ng/L at Klang West, commercial and cargo port. Coral reef Islands (Redang and Bidong) were relatively less contaminated compared to other coastal areas. The temporal variation revealed that only 1% of 28 stations sampled on November, 2011 was above the environmental risk limit of 24 ng/L as suggested by Dutch Authorities, while in January and April, 2012; 46% and 92% of the stations were above the limit respectively. The present findings demonstrate the wide detection of novel antifouling materials Irgarol-1051 which advocates the need for proper monitoring and conservation strategies for the coastal resources.