Merck
  • Management of differentiated thyroid cancer in the presence of resistance to thyroid hormone and TSH-secreting adenomas: a report of four cases and review of the literature.
Management of differentiated thyroid cancer in the presence of resistance to thyroid hormone and TSH-secreting adenomas: a report of four cases and review of the literature.
The Journal of clinical endocrinology and metabolism (2013-04-05)
Uğur Ünlütürk, Chutintorn Sriphrapradang, Murat Faik Erdoğan, Rifat Emral, Sibel Güldiken, Samuel Refetoff, Sevim Güllü
ABSTRACT
An increased or normal serum TSH concentration, despite elevated thyroid hormone levels, is observed in resistance to thyroid hormone (RTH) and TSH-secreting adenomas (TSHomas). When coexistent with a differentiated thyroid cancer (DTC), maintenance of a suppression of TSH is challenging. The aim of the study was to discuss the pitfalls arising from the failure to suppress TSH secretion in DTC and the strategies for proper treatment of DTC in association with RTH and TSHoma. Four unusual cases of DTC associated with TSHoma (2 cases), RTH (1 case), and an elevated TSH of unknown etiology (1 case) are presented, and the literature is reviewed. Although a persistent mild TSH elevation may not be a risk factor for the development of DTC, it represents an important problem during the treatment of DTC. Aggressive treatment options should be applied in the proper order to prevent tumor recurrence and persistence in the absence of ideal TSH suppression. Although there is no agreed consensus regarding the management of DTC in the presence of persistent hyperthyrotropinemia, complete tumor removal followed by radioablation and attempts to reduce the serum TSH to the lowest tolerable level are recommended. The outcomes in the reported cases have not been unfavorable, despite the persistence of nonsuppressed TSH.