Merck
  • Selective determination of hydroxyproline in urine by high-performance liquid chromatography using precolumn derivatization.
Selective determination of hydroxyproline in urine by high-performance liquid chromatography using precolumn derivatization.
Clinica chimica acta; international journal of clinical chemistry (1989-08-31)
T Teerlink, P Tavenier, J C Netelenbos
ABSTRACT
A method to measure total hydroxyproline in human urine was developed. Primary amino acids were derivatized with ortho-phthaldialdehyde, followed by derivatization of imino acids with 9-fluorenylmethyl chloroformate. The fluorescent 9-fluorenylmethyl chloroformate derivatives were separated by reversed phase high-performance liquid chromatography. 3,4-Dehydroproline was used as internal standard. Calibration curves for hydroxyproline and internal standard were linear from 1 to 200 pmol injected. Both within- and between-run precision were below 3.2%. Analytical recovery of hydroxyproline added to urine samples was 99.2 +/- 2.6%. Values for excreted hydroxyproline were determined by analysis of urine samples from adult volunteers. The hydroxyproline/creatinine ratio was found to be 15.8 +/- 4.6 mmol/mol (range 7.0-27.3) with no significant sex-related difference.