- Odor-associated idiopathic anaphylaxis. A case report.
Odor-associated idiopathic anaphylaxis. A case report.
Allergologia et immunopathologia (1995-01-01)
R L Saunders, G M Halpern, M E Gershwin
PMID7631593
ABSTRACT
A 44 year old woman is described who appears to have idiopathic anaphylaxis triggered by chemical odors. Her case and a general discussion of anaphylaxis are presented. The known causes of anaphylaxis and a discourse on idiopathic anaphylaxis are given. The treatment of idiopathic anaphylaxis is discussed.