- [Hygienic rationale for maximum permissible concentration of acetopropyl alcohol in atmospheric air].
[Hygienic rationale for maximum permissible concentration of acetopropyl alcohol in atmospheric air].
Gigiena i sanitariia (1993-09-01)
V V Neriueva, S M Novikov, N I Levchenko, N N Melńikova, M V Murkova, L N Semenovykh, T N Fursova, A P Pavlova, E E Kozeeva
PMID8026777
ABSTRACT
Toxic properties of acetopropyl alcohol were studied. Symptoms of CNS damages predominated in toxicodynamics. A maximal single concentration is 0.2 mg/m3 (4th class of danger).