Merck
  • Capillary affinophoresis of pea lectin with polyliganded affinophores: a model study of divalent-polyvalent interactions.
Capillary affinophoresis of pea lectin with polyliganded affinophores: a model study of divalent-polyvalent interactions.
Electrophoresis (1998-04-29)
K Shimura, K Kasai
ABSTRACT
Affinophoresis is a type of affinity electrophoresis using an affinophore, a soluble ionic carrier bearing affinity ligand(s). It was reported previously that an affinophore, prepared by coupling multiple p-aminophenyl alpha-D-mannoside ligands to a part of the carboxyl groups of succinylated polylysine, specifically changed the mobility of pea lectin in agarose gel. The affinophoresis of this divalent lectin with the polyliganded affinophore was investigated by using capillary electrophoresis. Analysis of the mobility change of the lectin in the presence of differently modified affinophores showed that the affinity was larger for affinophores having higher ligand density. Analysis of the inhibition of the mobility change by a neutral ligand, with a known affinity constant for the lectin, allowed estimation of the contributions of monovalent and divalent interactions to the binding in the lectin-affinophore complex. The proportion of divalent complexes was greater for affinophores having higher ligand density. This approach to estimate the contribution of divalency in complex formation should be generally applicable to the analysis of divalent interactions with different techniques other than electrophoresis.