Merck
  • HPLC determination and pharmacokinetics of thiabendazole and its major metabolite 5-OH thiabendazole in equine plasma.
HPLC determination and pharmacokinetics of thiabendazole and its major metabolite 5-OH thiabendazole in equine plasma.
Research in veterinary science (1987-07-01)
G E Hardee, M A Tshabalala, J N Moore, R D Gokhales
ABSTRACT
Separate high performance liquid chromatographic methods were developed for thiabendazole (TBZ) and 5-hydroxy thiabendazole (5-OH-TBZ) determination in horse plasma using 1-methyl-2-phenyl benzimidazole (MPBZ) as an internal standard. In both methods TBZ and 5-OH-TBZ were extracted from plasma using organic solvents, injected on to a C-18 column, and eluents monitored by a fluorescence detector. However, mobile phase composition, extraction solvent as well as detector wavelength differed in the two methods. The linear range for TBZ was 0.02 to 0.77 microgram ml-1 while that for 5-OH-TBZ was 0.96 to 8.0 micrograms ml-1. A commercially available TBZ oral suspension was administered to four thoroughbred horses in the following manner: days 1 and 2, 44 mg kg-1; days 4 and 5, 440 mg kg-1. Blood samples were collected during the 24 hours after administration and then analysed for TBZ and 5-OH-TBZ. Half-lives (t1/2), maximum plasma concentrations (Cmax), area under plasma concentration time curves (AUC O-alpha), and relative apparent bioavailability (F), were determined using pharmacokinetic equations. The pharmacokinetic parameters varied in the following manner: 1.16 to 13.63 hours (t1/2), 12 to 131 micrograms ml-1 X hours (AUC O-alpha), 3.33 to 8.90 micrograms ml-1 (Cmax), 1.38 to 0.12 (F) after 44 mg kg-1 and 440 mg kg-1 doses, respectively. The ratios of concentrations of TBZ to 5-OH-TBZ after oral administration of TBZ, were significantly lower for 44 mg kg-1 than 440 mg kg-1 doses.(ABSTRACT TRUNCATED AT 250 WORDS)