Merck
  • Reliable in vitro measurement of nitric oxide released from endothelial cells using low concentrations of the fluorescent probe 4,5-diaminofluorescein.
Reliable in vitro measurement of nitric oxide released from endothelial cells using low concentrations of the fluorescent probe 4,5-diaminofluorescein.
FEBS letters (2001-10-10)
J F Leikert, T R Räthel, C Müller, A M Vollmar, V M Dirsch
ABSTRACT
4,5-Diaminofluorescein (DAF-2) and its membrane-permeable derivate DAF-2 diacetate are fluorescent probes that have been developed to perform real-time biological detection of nitric oxide (NO). Their use for intracellular imaging, however, has recently been seriously questioned and data using DAF-2 for extracellular NO detection at low levels, as for example released from endothelial cells, are rare. Here we show that a reliable detection of low levels of NO in biological systems by DAF-2 is possible (a) by using low DAF-2 concentrations (0.1 microM) and (b) by subtracting the DAF-2 auto-fluorescence from the measured total fluorescence. The described method allows easy real-time detection of endothelial NO formation.