Merck
  • Peptic ulcer disease with related drug treatment in pregnant women and congenital abnormalities in their offspring.
Peptic ulcer disease with related drug treatment in pregnant women and congenital abnormalities in their offspring.
Congenital anomalies (2010-08-24)
Ferenc Bánhidy, Abdallah Dakhlaoui, Erzsébet H Puhó, Andrew E Czeizel
ABSTRACT
Peptic ulcer disease (PUD) is a common disease which can also occur in pregnant women. However, the possible association of PUD and related drug treatments in pregnant women with the risk of structural birth defects (i.e. congenital abnormalities [CA]) in their offspring has not been estimated in controlled population-based epidemiological studies. Thus, the prevalence of PUD in pregnant women who later delivered babies (cases) with different CA and in pregnant women who delivered newborns without CA (controls) was compared in the Hungarian Case-Control Surveillance of Congenital Abnormalities. Controls were matched to cases. Of 22,843 cases with congenital abnormalities, 182 (0.80%) had mothers with reported/recorded PUD, while of 38,151 controls, 261 (0.68%) were born to mothers with reported/recorded PUD. However, PUD(?) based on maternal information and/or unspecified diagnostic criteria, and PUD(!) based on endoscopic diagnosis showed different variables of mothers and newborn infants. Thus, finally, 20 case mothers and 58 control mothers with PUD(!) and related drugs were evaluated in detail. There was no higher risk for total CA group in the offspring of mothers with PUD during pregnancy (adjusted OR with 95% CI: 0.6, 0.3-0.9). Specific CA groups in cases were also assessed versus controls, but specified CA had no higher risk in the offspring of pregnant women with PUD and related drug treatments. In conclusion, a higher rate of CA was not found in the offspring of mothers with PUD.