Merck
  • Synthesis of 3-methoxyazetidines via an aziridine to azetidine rearrangement and theoretical rationalization of the reaction mechanism.
Synthesis of 3-methoxyazetidines via an aziridine to azetidine rearrangement and theoretical rationalization of the reaction mechanism.
The Journal of organic chemistry (2011-03-11)
Sonja Stanković, Saron Catak, Matthias D'hooghe, Hannelore Goossens, Kourosch Abbaspour Tehrani, Piet Bogaert, Michel Waroquier, Veronique Van Speybroeck, Norbert De Kimpe
ABSTRACT
The synthetic utility of N-alkylidene-(2,3-dibromo-2-methylpropyl)amines and N-(2,3-dibromo-2-methylpropylidene)benzylamines was demonstrated by the unexpected synthesis of 3-methoxy-3-methylazetidines upon treatment with sodium borohydride in methanol under reflux through a rare aziridine to azetidine rearrangement. These findings stand in contrast to the known reactivity of the closely related N-alkylidene-(2,3-dibromopropyl)amines, which are easily converted into 2-(bromomethyl)aziridines under the same reaction conditions. A thorough insight into the reaction mechanism was provided by both experimental study and theoretical rationalization.