Merck
  • Involvement of endoplasmic reticulum stress in isoliquiritigenin-induced SKOV-3 cell apoptosis.
Involvement of endoplasmic reticulum stress in isoliquiritigenin-induced SKOV-3 cell apoptosis.
Recent patents on anti-cancer drug discovery (2012-09-12)
Xuan Yuan, Bacui Yu, Yanming Wang, Jiangtao Jiang, Liangliang Liu, Hong Zhao, Wang Qi, Qiusheng Zheng
ABSTRACT
Isoliquiritigenin (ISL), a licorice chalconoid, is a bioactive agent with chemopreventive potential that has been patented for tumor treatment in China. This study investigated the mechanisms of ISL-induced apoptosis in ovarian carcinoma SKOV-3 cells. Cell viability was evaluated using a 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl-tetrazolium bromide assay. The apoptotic rate was determined via flow cytometry using an annexin V-FITC apoptosis detection kit. The intracellular reactive oxygen species (ROS) levels were assessed using a 2,7-dichlorofluorescein probe assay. Malondialdehyde (MDA) formation was determined via thiobarbituric acid reactive substance test. The expressions of growth arrest and DNA damage-inducible gene (GADD153/CHOP), 78 kDa glucose-regulated protein (GRP 78), α-subunit of eukaryotic initiation factor 2 (eIF2α) phosphorylation, activating transcription factor 6α (ATF6α), and unspliced form of X-box binding protein1 (XBP1U) were analyzed via Western blot. Caspase-3 and caspase-12 activities were assessed using a fluorometric kit. Findings indicate that ISL significantly inhibits SKOV-3 cell proliferation, increases intracellular ROS levels, and causes SKOV-3 cell apoptosis. Moreover, ISL-exposed SKOV-3 cells trigger endoplasmic reticulum (ER) stress, as indicated by the enhancement of ER stress-related molecules p-eIF2α, GADD153/CHOP, GRP78, XBP1 expression, and cleavage of ATF6α. However, caspase-12 inhibitor (Z-ATAD) effectively and partially prevents ROS and MDA formation and inhibits ISL-induced SKOV-3 cell apoptosis. ISL induces apoptosis via ER stress-triggered signaling pathways in SKOV-3 cells. ER stress-induced cancer cell apoptosis has been discussed in some patents.