- [Distribution and metabolism of azoxybenzene in the rat].
[Distribution and metabolism of azoxybenzene in the rat].
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete (1989-05-01)
M Kujawa, R Macholz, M Härtig, R Plass, B Nickel
PMID2750228
ABSTRACT
No sex differences could been shown after absorption and distribution of azoxybenzen in the rat. The highest level occur in the thyroid gland, the adrenals and in the fat tissues. The maximum level was reached 30 min. after application.