Merck
  • p38 mitogen-activated protein kinase-dependent and -independent intracellular signal transduction pathways leading to apoptosis in human neutrophils.
p38 mitogen-activated protein kinase-dependent and -independent intracellular signal transduction pathways leading to apoptosis in human neutrophils.
The Journal of biological chemistry (1998-05-09)
S C Frasch, J A Nick, V A Fadok, D L Bratton, G S Worthen, P M Henson
ABSTRACT
Human neutrophils undergo apoptosis spontaneously when cultured in vitro; however, the signal transduction pathways involved remain largely unknown. In some cell types, c-Jun NH2-terminal kinase and p38 mitogen-activated protein kinase (MAPK) have been implicated in the pathways leading to stress-induced apoptosis. In this study, we begin to define two pathways leading to apoptosis in the neutrophil induced either by stress stimuli (UV, hyperosmolarity, sphingosine) or by anti-Fas antibody or overnight culture in vitro (spontaneous apoptosis). Apoptosis induced by stress stimuli activated p38 MAPK, and apoptosis was inhibited by the specific p38 MAPK inhibitor, 6-(4-Fluorophenyl)-2.3-dihydro-5-(4-puridinyl)imidazo(2, 1-beta)thiazole dihydrochloride. Furthermore, differentiation of HL-60 cells toward the neutrophil phenotype resulted in a loss in c-Jun NH2-terminal kinase activation with concomitant acquisition of formylmethionylleucylphenylalanine-stimulatable and stress-inducible p38 MAPK activity as well as apoptosis blockade by the p38 MAPK inhibitor. In contrast, anti-Fas-induced or spontaneous apoptosis occurred independent of p38 MAPK activation and was not blocked by the inhibitor. Both pathways appear to utilize member(s) of the caspase family, since pretreatment with either Val-Ala-Asp-fluoromethyl ketone or Asp-Glu-Val-Asp-fluoromethyl ketone inhibited apoptosis induced by each of the stimuli. We propose the presence of at least two pathways leading to apoptosis in human neutrophils, a stress-activated pathway that is dependent on p38 MAPK activation and an anti-FAS/spontaneous pathway that is p38 MAPK-independent.