Merck
  • Huntingtin mediates dendritic transport of β-actin mRNA in rat neurons.
Huntingtin mediates dendritic transport of β-actin mRNA in rat neurons.
Scientific reports (2012-02-23)
Bin Ma, Jeffrey N Savas, Man-Shan Yu, Brady P Culver, Moses V Chao, Naoko Tanese
ABSTRACT
Transport of mRNAs to diverse neuronal locations via RNA granules serves an important function in regulating protein synthesis within restricted sub-cellular domains. We recently detected the Huntington's disease protein huntingtin (Htt) in dendritic RNA granules; however, the functional significance of this localization is not known. Here we report that Htt and the huntingtin-associated protein 1 (HAP1) are co-localized with the microtubule motor proteins, the KIF5A kinesin and dynein, during dendritic transport of β-actin mRNA. Live cell imaging demonstrated that β-actin mRNA is associated with Htt, HAP1, and dynein intermediate chain in cultured neurons. Reduction in the levels of Htt, HAP1, KIF5A, and dynein heavy chain by lentiviral-based shRNAs resulted in a reduction in the transport of β-actin mRNA. These findings support a role for Htt in participating in the mRNA transport machinery that also contains HAP1, KIF5A, and dynein.
Product Description
Human serum (cystatin C), ERM®, certified reference material
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8, ascites fluid, clone 1HU-4C8, Chemicon®
Pricing and availability is not currently available.