Merck
  • Concordance between Results of Medium-term Liver Carcinogenesis Bioassays and Long-term Findings for Carcinogenic 2-Nitropropane and Non-carcinogenic1-Nitropropane in F344 Rats.
Concordance between Results of Medium-term Liver Carcinogenesis Bioassays and Long-term Findings for Carcinogenic 2-Nitropropane and Non-carcinogenic1-Nitropropane in F344 Rats.
Journal of toxicologic pathology (2012-02-10)
Yuko Doi, Seiko Tamano, Mayumi Kawabe, Masashi Sano, Norio Imai, Hironao Nakashima, Fumio Furukawa, Akihiro Hagiwara, Masanori Otsuka, Tomoyuki Shirai
ABSTRACT
This study was conducted to determine the concordance of results for a pair of structural isomers, 2-nitropropane (2-NP) and 1-nitropropane (1-NP), using the rat medium-term liver carcinogenesis bioassay (Ito test) and previously published long-term carcinogenicity tests. Male F344 rats were given a single intraperitoneal injection of DEN (200 mg/kg b.w.) to initiate hepatocarcinogenesis. After 2 weeks, they received per os 0, 0.8, 4 or 20 mg/kg/day of 2-NP or 1-NP six times a week and were subjected to two-thirds partial hepatectomy at week 3. Non-initiated groups receiving 0 or 20 mg/kg/day were also included. The animals were sacrificed for quantitative analysis of GST-P-positive foci at week 8. With the highest dose of 2-NP, significantly increased numbers and areas of GST-P-positive foci were demonstrated as compared with the respective control but were not noted with 1-NP. In the non-DEN-initiated groups, many small GST-P-positive foci of less than 0.2 mm in diameter were also induced in the rats treated with 2-NP at 20 mg/kg/day but were lacking with 1-NP. These results strongly support that 2-NP is a complete hepatocarcinogen with a potent initiation activity, whereas 1-NP is not.