Merck
  • A digestion technique for the reduction of background staining in the immunoperoxidase method.
A digestion technique for the reduction of background staining in the immunoperoxidase method.
Journal of clinical pathology (1977-01-01)
M Reading
PMID65360