Merck
  • Vaso-occlusion in sickle cell disease: pathophysiology and novel targeted therapies.
Vaso-occlusion in sickle cell disease: pathophysiology and novel targeted therapies.
Blood (2013-09-21)
Deepa Manwani, Paul S Frenette
ABSTRACT
Recurrent and unpredictable episodes of vaso-occlusion are the hallmark of sickle cell disease. Symptomatic management and prevention of these events using the fetal hemoglobin-reactivating agent hydroxyurea are currently the mainstay of treatment. Discoveries over the past 2 decades have highlighted the important contributions of various cellular and soluble participants in the vaso-occlusive cascade. The role of these elements and the opportunities for therapeutic intervention are summarized in this review.