Merck
  • Confirmatory analysis of firocoxib in bovine milk by rapid resolution liquid chromatography tandem mass spectrometry.
Confirmatory analysis of firocoxib in bovine milk by rapid resolution liquid chromatography tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2009-01-31)
Geraldine Dowling, Pasquale Gallo, Liam Regan
ABSTRACT
A rapid method has been developed to analyse for firocoxib (FIRO) residue in bovine milk. Milk samples were extracted with acetonitrile and sample extracts were purified on Evolute ABN solid phase extraction cartridges. Aliquots were analysed by rapid resolution liquid chromatography tandem mass spectrometry (RRLC-MS/MS). The method was validated in bovine milk, according to the criteria defined in Commission Decision 2002/657/EC. The decision limit (CCalpha) was 1.18ng/mL and for the detection capability a (CCbeta) value of 2.02ng/mL was obtained. The measurement uncertainty of the method was 27%. Fortifying bovine milk samples (n=18) in three separate assays, show the accuracy of the method to be between 96 and 105%. The precision of the method, expressed as RSD values for the within-lab reproducibility at the three levels of fortification (5, 7.5 and 10ng/mL) was less than 11% respectively.