Merck
  • Effect of vitamin D analogues on acute cardiorenal syndrome: a laboratory rat model.
Effect of vitamin D analogues on acute cardiorenal syndrome: a laboratory rat model.
The Israel Medical Association journal : IMAJ (2014-02-12)
Ofer Havakuk, Michal Entin-Meer, Jeremy Ben-Shoshan, Pavel Goryainov, Sofia Maysel-Auslender, Erel Joffe, Gad Keren
ABSTRACT
Vitamin D has been shown to induce beneficial effects on cardiovascular and renal morbidity by regulating inflammation and tissue fibrosis. To evaluate the effect of vitamin D analogues on cardiac function and fibrosis in an animal model of cardiorenal syndrome. Unilateral nephrectomy was performed and myocardial infarction induced in rats. The rats were treated with vitamin D receptor activator (VDRA, paricalcitol, 40 ng/250 g x 3/week) versus a vehicle. A third group of animals, which served as the control, underwent sham surgery and received no treatment. After 4 weeks of treatment, cardiac function and fibrosis were assessed by trans-thoracic echo and histology, respectively. As a parameter of systemic inflammation, previously shown to be altered in acute coronary syndrome, T regulatory (Treg) cell levels were measured by flow cytometry. Renal dysfunction was documented by standard laboratory tests. After 4 weeks of treatment, no significant improvement in cardiac function parameters was noted following VDRA administration. VDRA treatment did not significantly alter Treg cell systemic levels. Consistently, despite a trend toward less extent of myocardial fibrosis, we found no clear beneficial effects of VDRA on myocardial tissue inflammation and remodeling. Vitamin D treatment showed no beneficial effects on cardiac function parameters and fibrosis in an animal model of cardiorenal syndrome.
Product Description
USP
Paricalcitol, United States Pharmacopeia (USP) Reference Standard