Merck
  • Gene structure and evolution of testicular haploid germ cell-specific genes, Oxct2a and Oxct2b.
Gene structure and evolution of testicular haploid germ cell-specific genes, Oxct2a and Oxct2b.
Genomics (2004-03-19)
Masayoshi Onishi, Teruo Yasunaga, Hiromitsu Tanaka, Yoshitake Nishimune, Masami Nozaki
ABSTRACT
OXCT/SCOT is the rate-determining enzyme in ketolysis in mitochondria of many extrahepatic organs. Two testicular isoforms, Oxct2a and Oxct2b, are highly homologous and specifically expressed in haploid spermatids of the mouse. In this report, we analyzed the structure and evolution of Oxct2a and Oxct2b. Both Oxct2's are single-copy intronless genes, of which nucleotide sequences are conserved with Oxct, indicating that these genes are transposons generated from Oxct. A CpG island was found within both Oxct2's. Oxct2a and Oxct2b are located in the third introns of Bmp8a and Bmp8b, and they are positioned within a 240-kb region in a tail-to-tail orientation on chromosome 4. This structural feature was also conserved in a syntenic region of human 1p34.3. Structural similarity between mice and humans indicated that these two sets of genes were generated by a segmental gene duplication, which occurred before the primate-rodent split. Dot matrix and phylogenetic tree analyses demonstrated that multiple rounds of intrachromosomal gene conversion between the two loci occurred in each species independently.