Merck
  • The use of non-tumor data in cancer risk assessment: reflections on butadiene, vinyl chloride, and benzene.
The use of non-tumor data in cancer risk assessment: reflections on butadiene, vinyl chloride, and benzene.
Regulatory toxicology and pharmacology : RTP (2003-03-29)
Richard Albertini, Harvey Clewell, Matthew W Himmelstein, Eric Morinello, Stephen Olin, Julian Preston, Louis Scarano, Martyn T Smith, James Swenberg, Raymond Tice, Curtis Travis
ABSTRACT
The estimation and characterization of a cancer risk is grounded in the observation of tumors in humans and/or experimental animals. Increasingly, however, other kinds of data (non-tumor data) are finding application in cancer risk assessment. Metabolism and kinetics, adduct formation, genetic damage, mode of action, and biomarkers of exposure, susceptibility, and effects are examples. While these and other parameters have been studied for many important chemicals over the past 30-40 years, their use in risk assessments is more recent, and new insights and opportunities are continuing to unfold. To provide some perspective on this field, the ILSI Risk Science Institute asked a select working group to characterize the pertinent non-tumor data available for 1,3-butadiene, benzene, and vinyl chloride and to comment on the utility of these data in characterizing cancer risks. This paper presents the findings of that working group and concludes with 15 simple principles for the use of non-tumor data in cancer risk assessment.
Product Description
Supelco
Vinyl chloride solution, certified reference material, 2000 μg/mL in methanol