- From miso, saké and shoyu to cosmetics: a century of science for kojic acid.
From miso, saké and shoyu to cosmetics: a century of science for kojic acid.
Natural product reports (2006-11-23)
Ronald Bentley
PMID17119644
ABSTRACT
This article reviews the curious history of kojic acid, discovered as a fungal natural product in 1907. It was one of the first secondary metabolites to have its biosynthetic pathway studied by the isotope tracer technique, and, more recently, has been of interest as a skin lightening agent. There are 112 references.