- A new method based on the use of diethyl pyrocarbonate as a nuclease inhibitor for the extraction of undegraded nucleic acid from plant tissues.
A new method based on the use of diethyl pyrocarbonate as a nuclease inhibitor for the extraction of undegraded nucleic acid from plant tissues.
European journal of biochemistry (1968-09-24)
F Solymosy, I Fedorcsák, A Gulyás, G L Farkas, L Ehrenberg
PMID5698613
MATERIALS