Merck
  • Enantioselective Friedel-Crafts alkylations catalyzed by bis(oxazolinyl)pyridine-scandium(III) triflate complexes.
Enantioselective Friedel-Crafts alkylations catalyzed by bis(oxazolinyl)pyridine-scandium(III) triflate complexes.
Journal of the American Chemical Society (2007-07-31)
David A Evans, Keith R Fandrick, Hyun-Ji Song, Karl A Scheidt, Risheng Xu
ABSTRACT
The enantioselective Friedel-Crafts addition of a variety of indoles catalyzed by bis(oxazolinyl)pyridine-scandium(III) triflate complexes (Sc(III)-pybox) was accomplished utilizing a series of beta-substituted alpha,beta-unsaturated phosphonates and alpha,beta-unsaturated 2-acyl imidazoles. The acyl phosphonate products were efficiently transformed into esters and amides, whereas the acyl imidazole adducts were converted to a broader spectrum of functionalities such as esters, amides, carboxylic acids, ketones, and aldehydes. The sense of stereoinduction and level of enantioselectivity were found to be functions of the size of the substrate employed, the substitution on the ligand, and the catalyst loading. Molecular modeling of the catalyst with the bound substrates was performed based on the crystal structures of the catalyst complexes and the sense of stereoinduction observed in the addition reaction. Nonlinear effects over a range of catalyst concentrations implicate a mononuclear complex as the active catalyst.