Merck
  • Preventive effects of rapamycin on inflammation and capillary degeneration in a rat model of NMDA-induced retinal injury.
Preventive effects of rapamycin on inflammation and capillary degeneration in a rat model of NMDA-induced retinal injury.
Biological & pharmaceutical bulletin (2015-03-10)
Yuto Aoki, Tsutomu Nakahara, Daiki Asano, Hiroko Ushikubo, Asami Mori, Kenji Sakamoto, Kunio Ishii
ABSTRACT
Inhibitors of the mammalian target of rapamycin (mTOR) have been shown to protect against neuronal injury, but the mechanisms underlying this effect are not fully understood. The present study aimed to examine the effects of rapamycin, an inhibitor of the mTOR pathway, on inflammation and capillary degeneration in a rat model of N-methyl-D-aspartate (NMDA)-induced retinal neurotoxicity. Inflammation and capillary degeneration were evaluated by counting the numbers of CD45-positive leukocytes and Iba1-positive microglia, and by measuring the length of empty basement membrane sleeves, respectively. Marked increases in the numbers of leukocytes and microglia were observed 1 d after intravitreal injection of NMDA (200 nmol), and significant capillary degeneration was observed after 7 d. These NMDA-induced changes were significantly reduced by the simultaneous injection of rapamycin (20 nmol) with NMDA. These results suggest that rapamycin has preventive effects on inflammation and capillary degeneration during retinal injury.
Product Description
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Pentobarbital, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%